missing translation for 'onlineSavingsMsg'
Learn More

Syringaldazine 99+%, Thermo Scientific Chemicals

Catalog Number p-7203416
Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
14414-32-5
C18H20N2O6
MYNVJNZGTAVRJD-UHFFFAOYSA-N
syringaldazine, 4-hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-dimethoxy-4-hydroxybenzalazine, 4,4'-hydrazine-1,2-diylidenebis methanylylidene bis 2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone, 4-1e-e-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, 4-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 2-4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone
5379425
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
14414-32-5
C18H20N2O6
MYNVJNZGTAVRJD-UHFFFAOYSA-N
syringaldazine, 4-hydroxy-3,5-dimethoxybenzaldehyde azine, 3,5-dimethoxy-4-hydroxybenzalazine, 4,4'-hydrazine-1,2-diylidenebis methanylylidene bis 2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone, 4-1e-e-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, 4-2-4-hydroxy-3,5-dimethoxyphenyl methylidene hydrazin-1-ylidene methyl-2,6-dimethoxyphenol, benzaldehyde, 4-hydroxy-3,5-dimethoxy-, 2-4-hydroxy-3,5-dimethoxyphenyl methylene hydrazone
5379425

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Materiale o nome chimico Syringaldazine
Nota nome 99+%
Punto di fusione 209°C to 213°C
Colore Green to Yellow
Spettro a infrarossi Authentic
Quantità 1 g
SMILES COC1=CC(=CNNC=C2C=C(C(=O)C(=C2)OC)OC)C=C(C1=O)OC
IUPAC Name 4-[[2-[(3,5-dimethoxy-4-oxocyclohexa-2,5-dien-1-ylidene)methyl]hydrazinyl]methylidene]-2,6-dimethoxycyclohexa-2,5-dien-1-one
Molecular Weight (g/mol) 360.366
Peso formulazione 360.36
Percent Purity ≥99.0%
Forma fisica Fine Crystalline Powder
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'