Learn More
Socorex™ Dosys™ All-glass Syringes
Codice prodotto. 10477072
Cambia vista
Click to view available options
Volume (sistema metrico):
0.1 to 1 mL
0.2 to 5 mL
0.5 to 2 mL
1 to 10 mL
1 to 20 mL
1 to 30 mL
1 to 50 mL
10 to 150 mL
10 to 200 mL
Tipo di collegamento:
Luer Lock
Luer Nozzle
Dimensione della confezione:
Pezzo
pezzi 2
pezzi 3
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Volume (sistema metrico) Tipo di collegamento unitSize
10477072 0.5 to 2 mL Luer Lock pezzi 3
10029761 1 to 20 mL Luer Lock pezzi 2
10131442 1 to 50 mL Luer Lock Pezzo
10039701 0.2 to 5 mL Luer Nozzle pezzi 3
10324932 1 to 20 mL Luer Nozzle pezzi 2
10161342 1 to 10 mL Luer Nozzle pezzi 3
10551713 0.2 to 5 mL Luer Lock pezzi 3
10385022 1 to 10 mL Luer Lock pezzi 3
11837253 0.1 to 1 mL Luer Nozzle pezzi 3
12663275 10 to 150 mL Luer Lock Pezzo
12673275 10 to 200 mL Luer Lock Pezzo
15562185 1 to 30 mL Luer Nozzle pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Codice prodotto. 10477072 Fornitore Socorex™ N. del fornitore 155.0502
Spedizione stimata: 27-08-2026
Aggiungi al carrello
Aggiungi al carrello
Resistant borosilicate glass syringes can be sterilized and reused. Socorex™ Dosys™ All-glass Syringes provide a low-cost alternative to disposable plastic syringes and are easy to disassemble for cleaning.
  • Borosilicate glass construction.
  • Superior chemical, heat and shock resistance.
  • Low-cost alternative to disposable plastic syringes.
  • Precision-machined plunger and barrel.
  • Excellent fitting and liquid tightness (not gas tight).
  • Permanent, high visibility graduations.
  • Glass Luer and metal Luer lock nozzles available.
  • Can be sterilized up to 160°C / 320°F.
  • Very easy to disassemble and clean.
Specifica
Autoclavabile Autoclavable
Materiale Glass, Glass (Barrel), Glass (Plunger), Metal (Tip)
Product Type Syringe
Graduazioni 0.1 mL
Quantità 3/Pk.
Volume (sistema metrico) 0.5 to 2 mL
Tipo di collegamento Luer Lock
Graduated Yes