Learn More
CELLPATH SYSTEM II+ BIOPSY CASSETTE
Product Code. 30226245
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226245 Threaded Pack of 2000
30226243 Loose Pack of 1000
30226244 Hoppered Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30226245 Supplier CELLPATH Supplier No. EAM060272B
  • ONLINE EXCLUSIVE PRICE
    307.37€
    245.96€ / pezzi 2000
    Risparmia 61.41€
    19% Sconto
 Restricted Item
Spedizione stimata: 14-12-2026
Incorporating 0.9 x 0.9 mm apertures, the System II+ Biopsy Cassette offers improved specimen security for small biopsy and tissue fragments. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Specifications
Color White
Material Threaded
Product Type Cassette