Learn More
CELLPATH SYSTEM II HEX - CASSETTE
Description
- The System II Hex Cassette benefits from an optimised writing surface as well as extra thick side walls that increase stability and rigidity, essential for high-quality sectioning. Supplied loose, threaded (for easy loading of Leica and Sakura printers) or pre-loaded in hopper tubes (for easy loading of Thermo Scientific cassette printers).
- Available in a range of colours, the System II Hex Cassette can be closed by using one of our plastic or stainless-steel lids.
- Key Benefits:
- Hex design maximises open surface area to increase the fluid exchange
- Optimised writing surface for printing high-quality text, 1D and 2D barcodes
- Writing surface suitable for marking with a CellMark marker pen and CellMark 2B pencil
- Large colour range available
Specifications
Tekniske data
| Color | White |
| Material | Laser Threaded |
| Product Type | Cassette |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?