Learn More
CELLPATH SYSTEM II+ ROUND CASSETTE
Product Code. 30226251
Change view
Click to view available options
Material:
Hoppered
Loose
Threaded
Unit Size:
Pack of 1000
Pack of 2000
Pack of 3000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
30226251 Threaded Pack of 2000
30226249 Loose Pack of 1000
30226250 Hoppered Pack of 3000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. Se returpolicy
Artikelnummer. 30226251 Leverandør CELLPATH Leverandørnr. EAM080272B
  • ONLINE EXCLUSIVE PRICE
    307.37€
    245.96€ / pezzi 2000
    Risparmia 61.41€
    19% Sconto
Se alle kampagner
 Restricted Item
Spedizione stimata: 16-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
The System II+ Round Cassette incorporates a round 2.4 mm diameter holes facilitates increased reagent flow in a traditional cassette.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Tekniske data
Color White
Material Threaded
Product Type Cassette
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.