Learn More
Invitrogen™ SYTO™ RNASelect™ Red
Descrizione
SYTO™ RNASelect™ Red is a cell-permeant, RNA-selective fluorescent dye for live or fixed eukaryotic cells. It allows researchers to visualize RNA-rich nucleoli and cytoplasmic RNA in a Texas Red/RFP compatible channel. Compared to SYTO™ RNASelect™ Green, it is compatible with 4% paraformaldehyde fixation rather than methanol fixation, and leaves GFP/FITC channels open for antibodies, fluorescent proteins, and other fluorescent probes in multiplex and high-content imaging.
This molecular probe is prepared as a ready-to-use 100x aqueous solution, appropriate for direct addition to either complete media or other buffers of your choice to then be used in staining cells.
Compatibility with 4% paraformaldehyde fixation and permeabilization expands the applicability of this RNA dye for immunofluorescence workflows, enabling its use in high content screening for nucleolar reorganization, cell stress assays, and deep learning computational models.
The staining patterns demonstrate high RNA specificity, with expected staining demonstrating bright nucleoli and diffuse ribosomal staining throughout the cytoplasm and is easily multiplexable with classic nuclear stains such as Hoechst 33342 or DAPI. This specificity is demonstrated by the sharp decrease in staining intensity upon treatment with RNase, all red signal is quickly depleted while the nuclear DNA dye staining remains intact.
Features
- Suitable for both live and fixed cell workflows, connecting live cell observation with endpoint imaging.
- Compatible with multiplex imaging outside of the Texas Red/RFP channels with various organelle markers such as Hoechst/DAPI, phalloidin, ER, lectins, or mitochondrial probes.
- Applicable to high-content imaging and RNA based nucleolar morphology studies, including workflows where nucleolar signal is used for segmentation or comparative phenotyping.
For researchers familiar with SYTO™ 14, SYTO™ RNASelect™ Red offers a red channel option for RNA associated morphology in Cell Painting related or other multiplex imaging workflows.
This dye complements SYTO™ RNASelect™ Green in channel planning and fixation choice: SYTO™ RNASelect™ Green remains the better fit when the green channel is desired and the sample type requires methanol fixation, whereas SYTO™ RNASelect™ Red is appropriate when a red RNA channel or 4% PFA workflow is preferred.
Available as a single vial as S32710 and in a 3-pack format as S32711.
Specifica
Specifica
| Colore | Red |
| Concentrazione | 1 mM in 25% DMSO/DI H2O |
| Content And Storage | •3 Vials •Store at 2°C to 30°C •Protect from light •Do not freeze |
| Range di eccitazione lunghezza d'onda | 595 nm |
| Tipo di colorante | Fluorogenic Organic Dye |
| Formato | Vial |
| Da utilizzare con (applicazione) | Live Cell Imaging, Fixed Cell Imaging, High Content Screening, Fluorescence Microscopy |
| Da utilizzare con (apparecchiatura) | Fluorescence Microscope, Spectral Imaging Instrument, Flow Cytometer, High Content Imager |
| N. di test | 900 Tests |
| Reattività | RNA, Nucleolar Structure |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.