missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descrizione
T-bet/TBX21 Monoclonal antibody specifically detects T-bet/TBX21 in Human samples. It is validated for Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry
Specifications
Specifications
| Antigene | T-bet/TBX21 |
| Applicazioni | Western Blot, Flow Cytometry (Intracellular Staining), Immunocytochemistry |
| Classificazione | Monoclonal |
| Clone | 525803 |
| Coniugato | DyLight 594 |
| Diluizione | Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry |
| Formulazione | 50mM Sodium Borate |
| Alias gene | T-bet, TBETT-PET, TBLYMT-cell-specific T-box transcription factor T-bet, T-box 21, T-box expressed in T cells, T-box protein 21, T-box transcription factor TBX21, Transcription factor TBLYM |
| Specie ospite | Mouse |
| Immunogeno | E. coli-derived recombinant human T-bet, Glu326-Asn535, Accession # Q9UL17 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?