missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAF1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 20260-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The antibody is specific to TAF1.
3-Phosphoglycerate dehydrogenase (PHGDH; EC 1.1.1.95) catalyzes the transition of 3-phosphoglycerate into 3-phosphohydroxypyruvate, which is the first and rate-limiting step in the phosphorylated pathway of serine biosynthesis, using NAD+/NADH as a cofactor.Specifica
TAF1 | |
Polyclonal | |
Unconjugated | |
TAF1 | |
BA2R, CCG1, CCGS, Cell cycle gene 1 protein, DYT3, KAT4, N TAF1, NSCL2, OF, P250, TAF(II)250, TAF1, TAF2A, TAFII 250, TAFII250, TBP associated factor 250 kDa | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
270627, 6872 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P21675, Q80UV9 | |
TAF1 | |
Peptide | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |