Learn More
Talin-1 Rabbit, anti-Human, Clone: 7D11, Proteintech
Codice prodotto. 18049815 Sfoglia Tutto Proteintech Prodotti
Cambia vista
Click to view available options
Quantità:
100 μL
20 μL
Dimensione della confezione:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18049815 100 μL 100µL
18068695 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 18049815 Fornitore Proteintech N. del fornitore 828564RR100UL
Rabbit Recombinant Monoclonal Antibody
The Talin-1 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human Talin-1. This antibody recognizes human, mouse, rat antigen. The Talin-1 antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifica
Antigene Talin-1
Applicazioni Western Blot, Immunofluorescence, ELISA
Clone 7D11
Concentrazione 1000 μg/mL
Simboli geni ILWEQ, KIAA1027, talin 1, TLN, TLN1
Specie ospite Rabbit
Quantità 100 μL
Primario o secondario Primary
ID gene (immissione) 7094
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Forma Liquid
Isotype IgG