Learn More
Tamoxifen, Tocris Bioscience™
Estrogen receptor partial agonist/antagonist
Marca: Tocris Bioscience 6342/100
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Tamoxifen is a selective estrogen receptor modulator (SERM). Exhibits antiestrogenic effects in mammary tissue and estrogen-like effects in bone. Anticancer. Tamoxifen citrate (Cat.No. 0999) is also available.Specifica
| 2-[4-[(1Z)-1,2-Diphenyl-1-buten-1-yl]phenoxy]-N,N-dimethylethanamine | |
| 10540-29-1 | |
| >99% | |
| 100 mg |