Learn More
Tanshinone IIA, MedChemExpress
Product Code. 16493836
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16493836 25 mg 25mg
16444767 50 mg 50mg
16428878 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16493836 Supplier Medchem Express Supplier No. HYN0135
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Tanshinone IIA (Tan IIA) is one of the main compositions in the root of red-rooted salvia. Tanshinone IIA may suppress angiogenesis by targeting the protein kinase domains of VEGF/VEGFR2.
Specifications
Chemical Name or Material Tanshinone IIA
CAS 568-72-9
Color Red
Purity Grade Notes Research
Molecular Formula C19H18O3
Quantity 25 mg
Synonym Dan Shen ketone
Solubility Information DMSO : 100 mg/mL (339.74 mM; Need ultrasonic)
SMILES O=C(C1=C2C=CC3=C1CCCC3(C)C)C(C4=C2OC=C4C)=O
Molecular Weight (g/mol) 294.34
Formula Weight 294.34
Percent Purity 99.17%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only