Learn More
Taxol, Tocris Bioscience™
Promotes assembly and inhibits disassembly of microtubules
Marca: Tocris Bioscience 1097/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Antitumor agent; promotes and stabilizes tubulin polymerization, causing cell cycle arrest. Induces autocatalytic activation of caspase-10 in CCRF-HSB-2 cells, triggering apoptosis.Specifica
| Taxol | |
| C47H51NO14 | |
| 50mg | |
| RCINICONZNJXQF-VAZQATRQSA-N | |
| (1S,2S,4S,7R,9S,10S,12R,15S)-4,12-bis(acetyloxy)-1,9-dihydroxy-15-{[(2R,3S)-2-hydroxy-3-phenyl-3-(phenylformamido)propanoyl]oxy}-10,14,17,17-tetramethyl-11-oxo-6-oxatetracyclo[11.3.1.0³,¹⁰.0⁴,⁷]heptadec-13-en-2-yl benzoate | |
| 133640187 | |
| >99% |
| 33069-62-4 | |
| MFCD00869953 | |
| paclitaxel, taxol, taxol a, abraxane, paxene, paxceed, plaxicel, yewtaxan, onxol, ebetaxel | |
| CC(=O)O[C@@H]1C2=C(C)[C@H](C[C@@](O)([C@@H](OC(=O)C3=CC=CC=C3)C3[C@@]4(CO[C@@H]4C[C@H](O)[C@@]3(C)C1=O)OC(C)=O)C2(C)C)OC(=O)[C@H](O)[C@@H](NC(=O)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 853.92 | |
| 853.92 |