missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TBX10 Polyclonal Antibody

Descrizione
This antibody detects endogenous protein at a molecular weight of 42 kDa. Purity is >95% by SDS-PAGE.
The T-box (Tbx) motif is present in a family of genes whose structural features and expression patterns support their involvement in developmental gene regulation. The Tbx gene family are largely conserved throughout metazoan evolution, and these genes code for putative transcription factors that share a uniquely defining DNA-binding domain. Tbx genes are a family of developmental regulators with more than 20 members recently identified in invertebrates and vertebrates. Mutations in Tbx genes are associated with the onset of several human diseases. Our understanding of functional mechanisms of Tbx products has come mainly from the prototypical T/Brachyury, which is a transcription activator.
Specifica
Specifica
| Antigene | TBX10 |
| Applicazioni | Immunohistochemistry (Paraffin), Western Blot |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TBX10 |
| N. accesso geni | O75333, Q810F8 |
| Alias gene | dancer; Dc; mmTBX7; T-box 10; T-box 10, pseudogene 1; T-box 13; T-box 7; T-box protein 10; T-box protein 13; T-box transcription factor TBX10; T-box-containing transcriptional activator; TBX10; Tbx10-ps1; Tbx13; TBX7 |
| Simboli geni | TBX10 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?