Learn More
Tocris Bioscience™ TC-S 7001
Potent and highly selective ROCK inhibitor; orally active
Marca: Tocris Bioscience™ 4961/1
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
TC-S 7001 (also known as BAY-549) is a potent and highly selective ROCK inhibitor (IC50 values are 0.6 and 1.1 nM for human ROCK1 and ROCK2, respectively), which exhibits >200-fold selectivity over TRK and FLT3 receptors, and >900-fold selectivity over a panel of other kinases and cardiovascular relevent enzymes and receptors. TC-S 7001 reduces blood pressure in normotensive and hypertensive rats, and is orally active.Specifica
| 6-Chloro-N4-[3,5-difluoro-4-[(3-methyl-1H-pyrrolo[2,3-b]pyridin-4-yl)oxy]phenyl]-2,4-pyrimidinediamine | |
| 867017-68-3 | |
| Azaindole-1 | |
| 1 mg |