missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TCF3/TCF4 Monoclonal Antibody (6F12-3)
GREENER_CHOICE

Codice prodotto. 11524932
Cambia vista
Click to view available options
Quantità:
25 μg
Dimensione della confezione:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11524932 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11524932 Fornitore Invitrogen™ N. del fornitore MA125418

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

TCF 4 (T cell factor 4) is a member of the TCF/LEF (lymphoid enhancer factor) family of transcription factors. Members of this family along with b catenin, are involved in mediating Wnt signaling by transactivating downstream genes. TCF 4 generates transcriptionally active complexes with b catenin in colon carcinoma cells. Expression of TCF 4 is highest in the CNS and intestinal epithelium where Wnt signaling also plays an essential role in development. Aberrant TCF 4 transcription activity has been implicated in carcinogenesis of intestinal and mammary epithlia.
TRUSTED_SUSTAINABILITY

Specifica

Antigene TCF3/TCF4
Applicazioni Immunohistochemistry (Paraffin), Western Blot
Classificazione Monoclonal
Clone 6F12-3
Concentrazione 0.53 mg/mL
Coniugato Unconjugated
Formulazione TBS with 30% glycerol and 0.08% sodium azide; pH 7.4
Gene Tcf4
N. accesso geni P15806, P15884, P15923, Q60722
Alias gene 5730422P05Rik; A1; AA408400; AGM8; ALF2; ASP-I2; AW209082; bHLHb19; BHLHB21; Class A helix-loop-helix transcription factor ME2; class B basic helix-loop-helix protein 19; Class B basic helix-loop-helix protein 21; E12; E12/E47; E2.2; E2-2; E2A; E47; FECD3; helix-loop-helix protein HE47; immunoglobulin enhancer-binding factor E12/E47; Immunoglobulin transcription factor 1; immunoglobulin transcription factor 2; immunoglobulin transcription factor-2; ITF1; ITF-1; ITF2; ITF-2; ITF-2b; kappa-E2-binding factor; Me2; MITF-2; MITF-2A; MITF-2B; negative vitamin D response element-binding protein; NOL1-TCF3 fusion; Pan1; Pan2; PTHS; SEF2; SEF-2; SEF2-1; SEF2-1A; SEF2-1B; SEF2-1D; SL3-3 enhancer factor 2; Tcf3; TCF-3; TCF4; TCF-4; Tcfe2a; TFE; Transcription factor 3; transcription factor 3 (E2A immunoglobulin enhancer binding factors E12/E47); transcription factor 3 variant 3; transcription factor 4; Transcription factor 4 (Immunoglobulin transcription factor 2) (ITF-2) (MITF-2) (SL3-3 enhancer factor 2) (SEF-2) (Class A helix-loop-helix transcription factor ME2); transcription factor 4, isoform C; transcription factor 4, isoform D; transcription factor 4, isoform E; transcription factor 4, isoform L; transcription factor 4, isoform M; transcription factor 4, isoform R; transcription factor A1; transcription factor E2a; Transcription factor E2-alpha; transcription factor ITF-1; VDIR; VDR interacting repressor; vitamin D receptor-interacting repressor
Simboli geni TCF3, Tcf4
Specie ospite Mouse
Immunogeno His-tagged fusion protein corresponding to amino acids 31-331 of human TCF.
Metodo di purificazione Protein G
Quantità 25 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 21413, 21423, 6925, 6929
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.