missing translation for 'onlineSavingsMsg'
Learn More

TCL1A Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marca:  Proteintech 10475-1-AP-150UL

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16896873

  • 325.38€ / 150µL

Spedizione stimata: 03-10-2022
per visualizzare lo stock disponibile
Aggiungi al carrello

Descrizione

Descrizione

Overexpression of the TCL1 gene in humans has been implicated in the development of mature T cell leukemia, in which chromosomal rearrangements bring the TCL1 gene in close proximity to the T-cell antigen receptor (TCR)-alpha or TCR-beta regulatory elements (summarized by Virgilio et al., 1998). In normal T cells TCL1 is expressed in CD4-/CD8- cells, but not in cells at later stages of differentiation. TCL1 functions as a coactivator of the cell survival kinase AKT (Laine et al., 2000).
Specifica

Specifica

TCL1A
Polyclonal
Unconjugated
TCL1A
Oncogene TCL 1, Oncogene TCL1, Protein p14 TCL1, T cell leukemia/lymphoma 1A, TCL1, TCL1A
Rabbit
Antigen Affinity Chromatography
RUO
8115
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P56279
TCL1A
TCL1A Fusion Protein Ag0786
150 μL
Primary
Human
Antibody
IgG
Videos
SDS
Documenti
Certificati
Offerte speciali

Offerte speciali