missing translation for 'onlineSavingsMsg'
Learn More

TCR alpha Antibody (34), PE/Atto594 - Chimeric, R&D Systems™

Product Code. 18602558 Shop All Bio Techne Products
Change view
Click to view available options
Quantità:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
18602558 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18602558 Supplier Novus Biologicals Supplier No. NBP252684PEATT594
 Articolo soggetto a restrizioni
Spedizione stimata: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

TCR alpha Monoclonal antibody specifically detects TCR alpha in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigene TCR alpha
Applicazioni Flow Cytometry
Classificazione Monoclonal
Clone 34
Coniugato PE/ATTO 594
Formulazione PBS
Alias gene C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
Specie ospite Rabbit
Immunogeno Purified recombinant soluble TCR from the T-cell hybridoma 85.33, corresponding to V alpha 11
Metodo di purificazione Protein A purified
Quantità 0.1 mL
Status giuridico RUO
Disciplina di ricerca Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
Primario o secondario Primary
ID gene (immissione) 6955
Target Species Human
Content And Storage Store at 4°C in the dark. Do not freeze.
Product Type Antibody
Forma Purified
Isotype IgG κ
Linea di prodotti Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.