Learn More
Techne™ 96-Well Sample Concentrator
Codice prodotto. 11503869
Cambia vista
Click to view available options
Temperature Range (Metric):
25°C to 200°C
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Temperature Range (Metric) Dimensione della confezione
11503869 25°C to 200°C Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11503869 Fornitore Techne™ N. del fornitore 3662041
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Quickly concentrate multiple samples in a block at once
  • Variable needle position suits various Dri-Block™ inserts
  • Needles, block, and block heater sold separately
Specifica
Temperature Range (Metric) 25°C to 200°C
Dimensioni (L x P x A) 295 x 240 x 530 mm