Learn More
Telotristat ethyl, MedChemExpress
Product Code. 16491687
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491687 50 mg 50mg
16478227 5 mg 5mg
16435518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16491687 Supplier Medchem Express Supplier No. HY13055A
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.
Telotristat ethyl (LX1032) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.
Specifications
Chemical Name or Material Telotristat ethyl
CAS 1033805-22-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C27H26ClF3N6O3
Quantity 50 mg
Synonym LX1032LX1606
Solubility Information DMSO : ≥ 101.5 mg/mL (176.53 mM)
SMILES N[C@@H](CC1=CC=C(C2=NC(N)=NC(O[C@H](C3=CC=C(Cl)C=C3N4N=C(C)C=C4)C(F)(F)F)=C2)C=C1)C(OCC)=O
Molecular Weight (g/mol) 574.98
Formula Weight 574.98
Percent Purity 99.12%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less
Research purposes only