missing translation for 'onlineSavingsMsg'
Learn More

Telotristat etiprate, MedChemExpress

Product Code. 16481687
Encompass_Preferred
Change view
Click to view available options
Quantità:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
16481687 50 mg 50mg
16468227 5 mg 5mg
16426096 100 mg 100mg
16425518 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16481687 Supplier Medchem Express Supplier No. HY13055
 Articolo soggetto a restrizioni
Spedizione stimata: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Telotristat etiprate (LX1606 Hippurate) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.

Telotristat etiprate (LX1606 Hippurate) is a novel, orally-delivered inhibitor of tryptophan hydroxylase that reduces serotonin production.

Specifications

Materiale o nome chimico Telotristat etiprate
CAS 1137608-69-5
Colore White
Note sul grado di purezza Research
Formula molecolare C36H35ClF3N7O6
Quantità 50 mg
Sinonimo LX1606 Hippurate
Informazioni di solubilità DMSO : ≥ 28 mg/mL (37.13 mM)
SMILES O=C([C@H](CC1=CC=C(C=C1)C2=CC(O[C@H](C3=C(C=C(C=C3)Cl)N4C=CC(C)=N4)C(F)(F)F)=NC(N)=N2)N)OCC.O=C(C5=CC=CC=C5)NCC(O)=O
Molecular Weight (g/mol) 754.15
Peso formulazione 754.15
Percent Purity 95.0%
Grado Research
Forma fisica Solid
Da utilizzare con (applicazione) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.