Learn More
TFA Dostmann Temperature/Humidity Transmitter
Codice prodotto. 17701218
Cambia vista
Click to view available options
Tipo:
Temperature/Humidity Transmitter
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Tipo Dimensione della confezione
17701218 Temperature/Humidity Transmitter Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17701218 Fornitore TFA Dostmann N. del fornitore 30.3180.IT
  • ONLINE EXCLUSIVE PRICE
    24.97€
    18.34€ / pezzi 1
    Risparmia 6.63€
    26% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 11-12-2026
Temperature transmitter for TFA wireless thermo-hygrometers and weather stations.
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3039, 30.3060
  • For wireless transmission of the outdoor climate
  • Display with temperature and humidity
  • Range: 100 m in open field
  • Update: every 4 seconds
  • Also for climate monitoring of remote rooms
Specifica
Tipo Temperature/Humidity Transmitter