Learn More
Testo™ Testoterm™ Temperature Strips
Codice prodotto. 10282161
Cambia vista
Click to view available options
Temperature Range (Metric):
37°C to 65°C
71°C to 110°C
Dimensione della confezione:
pezzi 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Temperature Range (Metric) Dimensione della confezione
10282161 71°C to 110°C pezzi 10
10028660 37°C to 65°C pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 10282161 Fornitore Testo™ N. del fornitore 0646 0916
  • ONLINE EXCLUSIVE PRICE
    49.15€
    32.00€ / pezzi 10
    Risparmia 17.15€
    34% Sconto
Spedizione stimata: 21-12-2026
Use these strips for monitoring the temperature of moving or small objects and long-term monitoring of transport and storage temperatures. Testoterm™ Temperature Strips can be applied to a variety of different surfaces and react to specific increases in temperature by changing color irreversibly within 2 to 3 seconds.
  • Self-adhesive temperature strips
  • Available in booklets of 10
  • Strips are peeled off and applied to the object you wish to measure
  • Change color when thresholds are exceeded within 2 to 3 seconds
Specifica
Montaggio Self-adhesive
Precisione (Temperatura) ±1.5°C
Temperature Range (Metric) 71°C to 110°C
Tipo Self-adhesive Temperature Strips