Learn More
TFA Dostmann Temperature Transmitter
Product Code. 12636967
Change view
Click to view available options
Type:
Temperature/Humidity Transmitter
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
12636967 Temperature/Humidity Transmitter Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12636967 Supplier TFA Dostmann Supplier No. 30.3181.IT
  • ONLINE EXCLUSIVE PRICE
    23.06€
    16.93€ / pezzi 1
    Risparmia 6.13€
    26% Sconto
 Restricted Item
Spedizione stimata: 11-12-2026
Temperature transmitter for TFA wireless thermo-hygrometers and weather stations.
Transmitter for TFA wireless thermo-hygrometers and weather stations: 30.3039, 30.3053, 30.3060
  • For wireless transmission of the outdoor temperature
  • With temperature display
  • With additional cable sensor
  • Update: every 4 seconds
  • Range: 100 m in open field
  • Also for temperature monitoring in remote rooms
Specifications
Type Temperature/Humidity Transmitter