Learn More
Soluzione tampone modello TEP 4 pH 4
Click to view available options
Quantità:
1000 ml
Dimensione della confezione:
1L
Descrizione
- pH buffers by WTW – precise and traceable to PTB/NIST in bulk size of 250 ml
- Life up to 3 years (unopened)
- Thrifty use
- Easiest handling by means of dosing device
- Technical buffering solutions for pH 4.01 is available in the practical 250 ml bottles
- All modern technical buffering solutions are traceable to PTB (Physical Technical Government Agency) or NIST (National Institute of Standards and Technology, USA)
Specifica
Specifica
| pH | 4 |
| Forma fisica | Liquido |
| Quantità | 1000 ml |
| Imballaggio | Flacone |
| Tracciabilità a NIST | Sì |