Learn More
Terazosin hydrochloride dihydrate, MedChemExpress
Product Code. 16438266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16438266 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16438266 Supplier Medchem Express Supplier No. HYB0371A
 Restricted Item
Spedizione stimata: 30-11-2026
MedChemExpress Terazosin hydrochloride dihydrate is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. Terazosin hydrochloride dihydrate works by relaxing blood vessels and the opening of the bladder. Terazosin hydrochloride dihydrate has the potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment.
Terazosin hydrochloride dihydrate is a quinazoline derivative and a competitive and orally active α1-adrenoceptor antagonist. Terazosin hydrochloride dihydrate works by relaxing blood vessels and the opening of the bladder. Terazosin hydrochloride dihydrate has the potential for benign prostatic hyperplasia (BPH) and high blood pressure treatment.
Specifications
Chemical Name or Material Terazosin hydrochloride dihydrate
CAS 70024-40-7
Color White
Purity Grade Notes Research
Molecular Formula C19H30ClN5O6
Quantity 100 mg
Solubility Information DMSO : 31.25 mg/mL (67.95 mM; Need ultrasonic) ∣H2O : 12.5 mg/mL (27.18 mM; Need ultrasonic)
SMILES O=C(N1CCN(C2=NC(N)=C3C=C(OC)C(OC)=CC3=N2)CC1)C4OCCC4.O.Cl.O
Molecular Weight (g/mol) 459.92
Formula Weight 459.92
Percent Purity 99.23%
Grade Research
Physical Form Solid
Show More Show Less
Research purposes only