Learn More
Terbinafine hydrochloride, Tocris Bioscience™
Selective TASK3 activator
Marca: Tocris Bioscience 6484/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Terbinafine hydrochloride is a selective TASK3 activator (pEC50 = 6.2). Exhibits >10-fold selectivity for TASK3 over TREK2, TRESK, THIK1 and TASK2. Also inhibits TWIK1 (pIC50 = 5.69). Also antifungal; inhibits fungal sterol biosynthesis by inhibition of squalene epoxidase.Specifica
| 78628-80-5 | |
| N-[(2E)-6,6-Dimethyl-2-hepten-4-yn-1-yl]-N-methyl-1-naphthalenemethanamine hydrochloride | |
| 50 mg |