missing translation for 'onlineSavingsMsg'
Learn More

Terbuthylazine metabolite CGA 324007 100 μg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer

Codice prodotto. 30308801
Cambia vista
Click to view available options
Quantità:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità unitSize
30308801 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308801 Supplier Dr. Ehrenstorfer Supplier No. DREA17305500MC100

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Materiale o nome chimico Terbuthylazine Metabolite CGA 324007
CAS 309923-18-0
Nomi componenti o analita Terbuthylazine Metabolite CGA 324007
Shipping Condition Room Temperature
Formula molecolare C7 H12 N4 O2
Concentrazione 100 μg/mL
Certificazione/Conformità ISO 17034
Sinonimo 1,3,5-Triazine-2,4(1H,3H)-dione, 6-[(1,1-dimethylethyl)amino]-, 6-[(1,1-Dimethylethyl)amino]-1,3,5-triazine-2,4(1H,3H)-dione
InChI Formula InChI=1S/C7H12N4O2/c1-7(2,3)11-4-8-5(12)10-6(13)9-4/h1-3H3,(H3,8,9,10,11,12,13)
SMILES CC(C)(C)Nc1nc(O)nc(O)n1
IUPAC Name 6-(tert-butylamino)-1,3,5-triazine-2,4-diol
Solvent or Matrix Acetonitrile/Methanol
Molecular Weight (g/mol) 184.1958
Quantità 1 mL
Peso formulazione 184.096 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.