Learn More
tert-Butoxycarbonyl-epsilon-aminocaproic Acid, TRC
Specifica
Specifica
| Chemical Name or Material | tert-Butoxycarbonyl-epsilon-aminocaproic Acid |
| CAS | 6404-29-1 |
| Molecular Formula | C11H21NO4 |
| Quantity | 10 g |
| Synonym | 6-[N-(tert-Butoxycarbonyl)amino]caproic Acid, BOC-epsilon-aminocaproic Acid, Boc-6-aminohexanoic Acid, N-(tert-Butoxycarbonyl)-6-aminocaproic Acid, N-(tert-Butoxycarbonyl)-6-aminohexanoic Acid, N-(tert-Butoxycarbonyl)-epsilon-aminocaproic Acid, N-(tert-Butoxycarbonyl)-epsilon-aminohexanoic Acid, N-(tert-Butyloxycarbonyl)-epsilon-aminocaproic Acid |
| InChI Formula | InChI=1S/C11H21NO4/c1-11(2,3)16-10(15)12-8-6-4-5-7-9(13)14/h4-8H2,1-3H3,(H,12,15)(H,13,14) |
| SMILES | CC(C)(C)OC(=O)NCCCCCC(=O)O |
| IUPAC Name | 6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
| Molecular Weight (g/mol) | 231.29 |
| Formula Weight | 231.1471 |