missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl 4-aminobutyrate hydrochloride, 97%, Thermo Scientific Chemicals

Catalog Number 15442669
41.33 EUR valido fino al 2024-03-29
Usa il codice promo "21615" per avere il tuo prezzo promozionale.


Click to view available options
Quantità:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
58640-01-0
C8H18ClNO2
195.687
MFCD00270592
CZVNLCOJFFYZPG-UHFFFAOYSA-N
tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu.hcl, h-gamma-abu-otbu hcl, gamma-aminobutyric acid t-butyl ester hcl, gamma-aminobutyric acid t-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, h-, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl, h-gamma-abu-otbu hydrochloride
24750478
tert-butyl 4-aminobutanoate;hydrochloride
CC(C)(C)OC(=O)CCCN.Cl
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
58640-01-0
C8H18ClNO2
195.687
MFCD00270592
CZVNLCOJFFYZPG-UHFFFAOYSA-N
tert-butyl 4-aminobutanoate hydrochloride, h-gamma-abu-otbu.hcl, h-gamma-abu-otbu hcl, gamma-aminobutyric acid t-butyl ester hcl, gamma-aminobutyric acid t-butyl ester hydrochloride, 4-aminobutyric acid tert-butyl ester hydrochloride, h-, a-abu-otbu.hcl, h-gaba-otbu . hcl, h-.gamma.-abu-otbu.hcl, h-gamma-abu-otbu hydrochloride
24750478
tert-butyl 4-aminobutanoate;hydrochloride
CC(C)(C)OC(=O)CCCN.Cl

Employed as an important intermediate for raw material for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Employed as an important intermediate for raw material for pharmaceutical.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico tert-Butyl 4-aminobutyrate hydrochloride
Colore White
Formula lineare H2NCH2CH2CH2CO2C(CH3)3·HCl
Peso formulazione 195.69
Percent Purity 97%
Quantità 1 g
Forma fisica Powder

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'