Learn More
tert-Butyl Acrylate (Stabilized with MEHQ), TRC
Specifica
Specifica
| Materiale o nome chimico | tert-Butyl Acrylate |
| CAS | 1663-39-4 |
| Formula molecolare | C7H12O2 |
| Quantità | 5 mL |
| Sinonimo | 1,1-Dimethylethyl 2-Propenoate, 2-Propenoic Acid tert-Butyl Ester, NSC 20950, tert-Butyl 2-Propenoate, tert-Butyl Acrylate, tert-Butyl Propenoate |
| InChI Formula | InChI=1S/C7H12O2/c1-5-6(8)9-7(2,3)4/h5H,1H2,2-4H3 |
| SMILES | CC(C)(C)OC(=O)C=C |
| IUPAC Name | tert-butyl prop-2-enoate |
| Molecular Weight (g/mol) | 128.17 |
| Peso formulazione | 128.0837 |