missing translation for 'onlineSavingsMsg'
Learn More

tert-Butyl bromoacetate, 98%, Thermo Scientific Chemicals

Catalog Number p-7022238
Click to view available options
Quantità:
10 g
50 g
250 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5292-43-3
C6H11BrO2
195.06
MFCD00000188
BNWCETAHAJSBFG-UHFFFAOYSA-N
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat
79177
tert-butyl 2-bromoacetate
CC(C)(C)OC(=O)CBr
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
5292-43-3
C6H11BrO2
195.06
MFCD00000188
BNWCETAHAJSBFG-UHFFFAOYSA-N
tert-butyl bromoacetate, t-butyl bromoacetate, acetic acid, bromo-, 1,1-dimethylethyl ester, bromoacetic acid tert-butyl ester, tert-butylbromoacetate, acetic acid, bromo-, tert-butyl ester, brch2c o oc ch3 3, acetic acid, 2-bromo-, 1,1-dimethylethyl ester, bromo-acetic acid tert-butyl ester, tert-butylbromacetat
79177
tert-butyl 2-bromoacetate
CC(C)(C)OC(=O)CBr

tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
tert-Butyl bromoacetate is used as an intermediate in the production of dyes, pharmaceuticals, agrochemicals and in organic compounds. It finds application as an alkylating agent. It plays an important role to prepare (2-oxo-4-vinyl-azetidin-1-yl)-acetic acid tert-butyl ester and in collagenase inhibitors synthesis.

Solubility
Miscible with ethanol, chloroform and ethyl acetate. Immiscible with water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico tert-Butyl bromoacetate
Densità 1.33
Punti di ebollizione 175°C
Punto d'infiammabilità 49°C (120°F)
Odore Pungent
Formula lineare BrCH2CO2C(CH3)3
Indice di rifrazione 1.445
Quantità 10 g
Numero UN UN2924
Beilstein 1753010
Informazioni di solubilità Miscible with ethanol,chloroform and ethyl acetate. Immiscible with water.
Peso formulazione 195.06
Percent Purity 98%
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'