Learn More
(tert-Butyldimethylsilyloxy)acetaldehyde, TRC
Specifica
Specifica
| Materiale o nome chimico | (tert-Butyldimethylsilyloxy)acetaldehyde |
| CAS | 102191-92-4 |
| Formula molecolare | C8H18O2Si |
| Quantità | 2.5 g |
| Sinonimo | Dimethyl-tert-butylsilyloxyacetaldehyde, (tert-Butyldimethylsilanyloxy)acetaldehyde, 2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]acetaldehyde |
| InChI Formula | InChI=1S/C8H18O2Si/c1-8(2,3)11(4,5)10-7-6-9/h6H,7H2,1-5H3 |
| SMILES | O=CCO[Si](C)(C)C(C)(C)C |
| IUPAC Name | 2-((tert-butyldimethylsilyl)oxy)acetaldehyde |
| Molecular Weight (g/mol) | 174.31 |
| Peso formulazione | 174.11 |