missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals TES, 0.2M buffer soln., pH 7.5
Click to view available options
Quantità:
100 mL
250 mL
Descrizione
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.
Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TES, 0.2M buffer soln., is a suitable buffer for epidermal cell growth at pH 7.4 - 7.9, and is also useful for studying succinate oxidation. It is also widely used as a buffer in biological applications.
Solubility
Soluble in water (350 g/L) at 20°C, and methanol(very faint turbidity).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifica
Materiale o nome chimico | TES buffer soln. |
Concentrazione | 0.2 M |
pH | 7.5 |
Forma fisica | Liquid |
Quantità | 100 mL |
Informazioni di solubilità | Soluble in water (350g/L) at 20°C,and methanol(very faint turbidity). |
IUPAC Name | sodium;2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'