Learn More
Fisherbrand™ Test Tubes, Borosilicate Glass 3.3, Without Rim, Medium Wall
Codice prodotto. 17991122 Sfoglia Tutto Fisherbrand Traceable Prodotti
Cambia vista
Click to view available options
Capacità (sistema metrico):
5 mL
12 mL
13 mL
17 mL
21 mL
27 mL
32 mL
34 mL
53 mL
57 mL
112 mL
Lunghezza (sistema metrico):
75 mm
100 mm
125 mm
130 mm
150 mm
150 mm
160 mm
180 mm
200 mm
Diametro (sistema metrico):
12 mm
14 mm
16 mm
18 mm
20 mm
24 mm
25 mm
30 mm
Dimensione della confezione:
pezzi 100
pezzi 50
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) Lunghezza (sistema metrico) Diametro (sistema metrico) Dimensione della confezione
17991122 13 mL 100 mm 16 mm pezzi 100
17981142 5 mL 75 mm 12 mm pezzi 100
17901142 12 mL 130 mm 14 mm pezzi 100
15822295 17 mL 125 mm 16 mm pezzi 100
17951122 17 mL 130 mm 16 mm pezzi 100
15802285 21 mL 150 mm 16 mm pezzi 100
17931122 21 mL 160 mm 16 mm pezzi 100
17961132 27 mL 150 mm 18 mm pezzi 100
17921142 32 mL 180 mm 18 mm pezzi 100
17941122 34 mL 150 mm 20 mm pezzi 100
17941152 53 mL 150 mm 24 mm pezzi 50
17931142 57 mL 150 mm 25 mm pezzi 50
17911142 112 mL 200 mm 30 mm pezzi 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Codice prodotto. 17991122
Spedizione stimata: 15-12-2026
Aggiungi al carrello
Aggiungi al carrello
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from borosilicate glass 3.3
  • Thick-walled and therefore mechanically very resistant
  • With round bottom
Specifica
Capacità (sistema metrico) 13 mL
Lunghezza (sistema metrico) 100 mm
Materiale Borosilicate Glass (3.3)
Forma Cylindrical
Sterilità Non-sterile
Autoclavabile Autoclavable
Graduated No
Diametro (sistema metrico) 16 mm
Product Type Test Tube
Spessore della parete 1.2 mm