missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium iodide, 98%, Thermo Scientific Chemicals

Catalog Number p-7022619
Click to view available options
Quantità:
50 g
250 g
1000 g
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
311-28-4
C16H36IN
369.38
MFCD00011636
DPKBAXPHAYBPRL-UHFFFAOYSA-M
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
67553
tetrabutylazanium;iodide
[I-].CCCC[N+](CCCC)(CCCC)CCCC
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
311-28-4
C16H36IN
369.38
MFCD00011636
DPKBAXPHAYBPRL-UHFFFAOYSA-M
tetrabutylammonium iodide, tetra-n-butylammonium iodide, tbai, tetrabutylazanium iodide, tetra-n-butylammoniumjodid, tetrabutyl ammonium iodide, 1-butanaminium, n,n,n-tributyl-, iodide, n,n,n-tributyl-1-butanaminium iodide, tetra-n-butylammoniumjodid czech, tetrabutylammonium ion iodide
67553
tetrabutylazanium;iodide
[I-].CCCC[N+](CCCC)(CCCC)CCCC

Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetra-n-butylammonium iodide is used as a catalyst for methylation and benzylation of alcohols with dimethyl sulfate and benzyl bromide respectively. It is a useful source of iodide anion and involved in the preparation of neopentyl iodides from the triflates. It is also used in the preparation of novel quaternary amines, which serve as an antibacterial agent in the rise of drug-resistant bacteria. It acts as a phosphonium reversible inhibitors of cholinesterases. It is associated with boron trifluoride diethyl¬ etherate and used in the cleavage of ethers.

Solubility
Soluble in water and methanol. Insoluble in benzene.

Notes
Light sensitive and hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Materiale o nome chimico Tetra-n-butylammonium iodide
Punto di fusione 142°C to 148°C
Quantità 50 g
Beilstein 3916152
Sensibilità Hygroscopic; Light sensitive
Informazioni di solubilità Soluble in water and methanol. Insoluble in benzene.
Peso formulazione 369.38
Percent Purity 98%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'