Learn More
Tetrabutylphosphonium 2H,2H-Perfluorodecanoate, TRC
Codice prodotto. 30135261
Cambia vista
Click to view available options
Quantità:
10 mg
50 mg
100 mg
Dimensione della confezione:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
30135261 50 mg 50mg
30135073 100 mg 100mg
17982325 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Codice prodotto. 30135261 Fornitore Toronto Research Chemicals N. del fornitore TRCT31065550MG
 Articolo soggetto a restrizioni
Spedizione stimata: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 882489-14-7
Formula molecolare C16 H36 P . C10 H2 F17 O2
Molecular Weight (g/mol) 736.53
Sinonimo Phosphonium, tetrabutyl-, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoate (1:1), Phosphonium, tetrabutyl-, salt with 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoic acid (1:1) (9CI), Tetrabutylphosphonium 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecanoate, 8:2 FTCA tetrabutylphosphonium salt, 8:2 Fluorotelomer carboxylic acid tetrabutylphosphonium salt, Perfluoro-octylethanoic acid tetrabutylphosphonium salt, Tetrabutylphosphonium 2H,2H-perfluorodecanoate
IUPAC Name 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoate;tetrabutylphosphanium
SMILES CCCC[P+](CCCC)(CCCC)CCCC.[O-]C(=O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Specifica
Materiale o nome chimico Tetrabutylphosphonium 2H,2H-Perfluorodecanoate
Quantità 50 mg
InChI Formula InChI=1S/C16H36P.C10H3F17O2/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;11-3(12,1-2(28)29)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h5-16H2,1-4H3;1H2,(H,28,29)/q+1;/p-1
Peso formulazione 736.2338