Learn More
Tetramethylammonium Tetrafluoroborate, TRC
Specifica
Specifica
| Materiale o nome chimico | Tetramethylammonium Tetrafluoroborate |
| CAS | 661-36-9 |
| Formula molecolare | C4H12BF4N |
| Quantità | 2.5 g |
| Sinonimo | Methanaminium','N,N,N-trimethyl-','tetrafluoroborate(1-) (1:1) (ACI), Ammonium compounds','substituted','tetramethyl-','tetrafluoroborate (7CI), Ammonium','tetramethyl-','tetrafluoroborate(1-) (8CI), Borate(1-)','tetrafluoro-','N,N,N-trimethylmethanaminium (ZCI), Borate(1-)','tetrafluoro-','tetramethylammonium (8CI), Methanaminium','N,N,N-trimethyl-','tetrafluoroborate(1-) (9CI), Tetramethylammonium fluoroborate (6CI), [Me4N][BF4], Tetramethylammonium tetrafluoroborate, Tetramethylammonium tetrafluoroborate(1-) |
| InChI Formula | InChI=1S/C4H12N.BF4/c1-5(2,3)4;2-1(3,4)5/h1-4H3;/q+1;-1 |
| SMILES | C[N+](C)(C)C.F[B-](F)(F)F |
| IUPAC Name | tetramethylazanium;tetrafluoroborate |
| Molecular Weight (g/mol) | 160.95 |
| Peso formulazione | 161.0999 |