Learn More
Tetraxolin, TRC
Product Code. 30138107
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30138107 500 mg 500mg
30134987 250 mg 250mg
17902505 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30138107 Supplier Toronto Research Chemicals Supplier No. TRCN756885500MG
 Restricted Item
Spedizione stimata: 11-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 100343-43-9
Molecular Formula C10 H8 O6
Molecular Weight (g/mol) 224.167
Synonym 2,3-Dihydro-2,2,3,3-tetrahydroxy-1,4-naphthalenedione;2,2,3,3-Tetrahydroxynaphthalene-1,4-dione;Oksolin, Oxolin, Oxoline;
IUPAC Name 2,2,3,3-tetrahydroxytetralin-1,4-dione
SMILES OC1(O)C(=O)c2ccccc2C(=O)C1(O)O
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C10H8O6/c11-7-5-3-1-2-4-6(5)8(12)10(15,16)9(7,13)14/h1-4,13-16H
Formula Weight 224.0321
Chemical Name or Material Tetraxolin