missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TFAP2D Polyclonal Antibody

Descrizione
Antibody detects endogenous levels of total TFAP2D.
Sequence-specific DNA-binding protein that interacts with inducible viral and cellular enhancer elements to regulate transcription of selected genes. AP-2 factors bind to the consensus sequence 5'-GCCNNNGGC-3' and activate genes involved in a large spectrum of important biological functions including proper eye, face, body wall, limb and neural tube development. They also suppress a number of genes including MCAM/MUC18, C/EBP alpha and MYC.
Specifica
Specifica
| Antigene | TFAP2D |
| Applicazioni | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classificazione | Polyclonal |
| Concentrazione | 1 mg/mL |
| Coniugato | Unconjugated |
| Formulazione | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TFAP2D |
| N. accesso geni | Q7Z6R9, Q91ZK0 |
| Alias gene | activating enhancer binding protein 2 beta-like 1; activating enhancer-binding protein 2 delta; activating enhancer-binding protein 2-delta; AP-2 like; AP-2-delta; AP2-delta; hypothetical protein LOC566838; si:dkey-5e16.2; Tcfap2d; TFAP2BL1; TFAP2D; transcription factor AP-2 beta (activating enhancer binding protein 2 beta)-like 1; transcription factor AP-2 beta (activating enhancer-binding protein 2 beta)-like 1; transcription factor AP-2 beta-like 1; transcription factor AP-2 delta; transcription factor AP-2 delta (activating enhancer binding protein 2 delta); transcription factor AP-2, delta; transcription factor AP-2-beta-like 1; Transcription factor AP-2-delta; zgc:114196 |
| Simboli geni | TFAP2D |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?