missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFDP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11043-1-AP-150UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
DP family contains at least two members, DP1 and 2. Structurally, DP1 and 2 are relatively conserved, yet their expression appears to be tissue specific. E2F 1, a functional target for the tumor suppressor protein Rb, heterodimerizes with DP1 and forms an active E2F transcriptional complex. The interaction between members of the E2F and the DP families is mediated, in part, by a leucine repeat which is weakly conserved between the two families.Specifica
TFDP1 | |
Polyclonal | |
Unconjugated | |
TFDP1 | |
Dp 1, DP1, DRTF1, DRTF1 polypeptide 1, E2F dimerization partner 1, TFDP1, transcription factor Dp 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
21781, 361178, 7027 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q08639, Q14186 | |
Tfdp1 | |
TFDP1 Fusion Protein Ag1511 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |