Learn More
Theophylline-7-acetic Acid, TRC
Product Code. 30008562
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008562 1 g 1g
30138116 10 g 10g
30136214 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30008562 Supplier Toronto Research Chemicals Supplier No. TRCT3438601G
 Restricted Item
Spedizione stimata: 02-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 652-37-9
Molecular Formula C9 H10 N4 O4
Molecular Weight (g/mol) 238.2
Synonym 7H-Purine-7-acetic acid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo-Purine-7-acetic acid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo- (6CI,7CI,8CI), 1,2,3,6-Tetrahydro-1,3-dimethyl-2,6-dioxopurine-7-acetic acid, 7-(Carboxymethyl)theophylline, 7-Theophyllineacetic acid, 7-Theophyllinylacetic acid, Acefylline, Acephylline, Carboxymethyltheophylline, N-7-Theophyllineacetic acid, NSC 52996, Theophyllineacetic acid
IUPAC Name 2-(1,3-dimethyl-2,6-dioxopurin-7-yl)acetic acid
SMILES CN1C(=O)N(C)c2ncn(CC(=O)O)c2C1=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C9H10N4O4/c1-11-7-6(8(16)12(2)9(11)17)13(4-10-7)3-5(14)15/h4H,3H2,1-2H3,(H,14,15)
Formula Weight 238.0702
Chemical Name or Material Acefylline