Learn More
Thermo Scientific™ BetaSil™ C18 Javelin™ Guard Columns
Product Code. 11553231 Shop All Thermo Scientific Products
Change view
Click to view available options
Diameter (Metric) Inner:
2.1 mm
Length (Metric):
10 mm
20 mm
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Inner Length (Metric) Unit Size
11553231 2.1 mm 20 mm Pack of 4
11548720 2.1 mm 10 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11553231 Supplier Thermo Scientific™ Supplier No. 70105022106
 Restricted Item
Spedizione stimata: 27-11-2026
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
Features of Betasil C18 Solid Phase
  • Ligand on 100Å silica
  • High surface area with high bonded phase coverage
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifications
Particle Size 3 μm
Length (Metric) 20 mm
Diameter (Metric) Inner 2.1 mm
Stationary Phase C18
Product Line BetaSil
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.