Learn More
Epredia™ Disposable Blade Carriers
Product Code. 12063119 Shop All Epredia Products
Change view
Click to view available options
For Use With (Equipment):
Long Stroke Units
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
12063119 Long Stroke Units Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12063119 Supplier Epredia™ Supplier No. 705830
  • ONLINE EXCLUSIVE PRICE
    1920.00€
    1623.12€ / pezzi 1
    Risparmia 296.88€
    15% Sconto
In stock
Utilize a quick clamp system that accepts low- and high-profile blades
Updated models (long stroke and version 2 bar guideways) include removable angle adjustment and blade clamp levers.
Updated Disposable Blade Carriers
  • Blade angle and blade adjustment levers are removable to help protect user settings
  • Concentric lever locks onto the dovetail to help reduce accidental movement of the holder
  • Only compatible with newer HM 325, HM 355S, and HM 340E models which have serial numbers above 34000
Standard Disposable Blade Carriers
  • Quick clamp system for high- and low-profile blades
  • Clearance angle is adjustable
  • Upper part moves laterally without opening the blade clamp
  • Moveable knife guard covers the blade entirely in any position
  • Complete with knife carrier base
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Long Stroke Units