Learn More
Thermo Scientific™ GENESYS™ 20 Spectrophotometer Filter Holder
Marca: Thermo Scientific™ 4028000
Dettagli aggiuntivi : Peso : 0.80000kg
Descrizione
Filter Holder. Holds one filter/lens up to 8mm thick with dimensions up to 44mm wide and 104mm high.
Specifica
Filter Holder | |
Holds one filter/lens up to 8mm thick with dimensions 104 H x 44mm W | |
Genesys™ 20 visible spectrophotometer |