Learn More
Thermo Scientific™ HAAKE™ Viscotester™ iQ Rheometer Application Packages
Ready to use in seconds in any configuration you need
Marca:  Thermo Scientific™ 262-0010
Dettagli aggiuntivi : Peso : 14.00000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 15335123
  • 19920.00€ / Pezzo
Spedizione stimata
12-03-2019
Aggiungi al carrello
Descrizione
Descrizione
Acquire simple viscosity curves or perform complex rheological investigations covering measurements of low viscous liquids up to highly viscous pastes with the Thermo Scientific™ HAAKE™ Viscotester™ rheometer, which can be used as a standalone unit or optionally, be controlled by software. The size and footprint of the HAAKE™ Viscotester™ rheometer make it a portable unit suitable for mobile use or allow it to serve as an important tool in the quality control laboratory.
Application packages are designed to perform a diverse set of rheological tasks and meet a variety of application needs.
  • Advanced design with one-hand lift operation
  • Touchscreen control panel with multilingual, graphical user interface
  • Data quality assessment and QC quality criteria
  • Connect Assist quick coupling and automatic recognition of temperature modules and rotors
  • Liquid temperature module uses a circulator for coaxial cylinders and parallel plates
  • Self-sufficient Peltier temperature module for coaxial cylinders and parallel plates
  • Small dimensioned, coaxial cylinders use less sample and shorter temperature equilibration times to minimize costs per measurement
  • Universal container holder for relative measurements in the original containers
Specifica
Specifica
Food, paints (like printing pastes), polymers like resins, stablizer and thickening agents, cosmetics
HAAKE Viscotester iQ Rheometer, Peltier Temperature Module TM-PE-C with Heat Exchanger HXiQ, Cup CCB25 DIN, Rotor CCB25 DIN/Ti, Measuring Plate Insert, Gap Setting tool, 1.0mm caliper, Lower plate TMP35, Rotor P35/Ti
Application Package for Liquids and Pastes
Videos
Documentazione
Documentazione tecnica