Learn More
Thermo Scientific™ Hypersil GOLD™ aQ Javelin™ Guard HPLC Columns
Codice prodotto. 11568690 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Dimensione particelle:
5 μm
Dimensione della confezione:
pezzi 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Dimensione particelle Dimensione della confezione
11568690 5 μm pezzi 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 11568690 Fornitore Thermo Scientific™ N. del fornitore 25305012106
Spedizione stimata: 15-12-2026
Javelin guard columns offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, patented fingertight design has a 1/16 in. CPI tip that attaches directly to the inlet of the HPLC column with tubing or wrenches
  • Requires no holder, allowing quick and easy replacement
Hypersil GOLD aQ columns provide enhanced retention and resolution of polar analytes.
  • Polar endcapped C18 phase for alternative selectivity
  • Retention and resolution of polar analytes
  • Ideal for highly aqueous mobile phase
  • Excellent peak shape
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your local Customer Service for more information.
TRUSTED_SUSTAINABILITY
Specifica
Dimensione particelle 5 μm
Lunghezza (sistema metrico) 10 mm
Diametro (sistema metrico) interno 2.1 mm
Fase stazionaria aQ
Product Line Hypersil GOLD