Learn More
Thermo Scientific™ Precolonne Hypersil GOLD™ Javelin™
Codice prodotto. 11588680 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Diametro (sistema metrico) interno:
2.1 mm
4 mm
Lunghezza (sistema metrico):
10 mm
Dimensione particelle:
3 μm
5 μm
Dimensione della confezione:
pezzi 4
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Diametro (sistema metrico) interno Lunghezza (sistema metrico) Dimensione particelle unitSize
11588680 2.1 mm 10 mm 5 μm pezzi 4
11593121 4 mm 10 mm 3 μm pezzi 4
11583121 2.1 mm 10 mm 3 μm pezzi 4
11598680 4 mm 10 mm 5 μm pezzi 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Codice prodotto. 11588680 Fornitore Thermo Scientific™ N. del fornitore 25005012106
Spedizione stimata: 07-08-2026
Aggiungi al carrello
Aggiungi al carrello
Offer convenient, easy to use protection for analytical HPLC columns
  • Direct-connection, fingertight design for convenience and maximum efficiency
  • Patented design requires no holder, allowing quick and easy replacement
  • Hypersil GOLD HPLC columns provide excellent peak shape for all analyte types
  • Significant reduction in peak tailing while retaining C18 selectivity
  • Excellent resolution, efficiency and sensitvity
  • Confidence in the accuracy and quality of analytical data
Order Info
Javelin guards are available in other Thermo Scientific phases. Please call your Customer Service Representative for more information.
TRUSTED_SUSTAINABILITY
Specifica
Dimensione particelle 5 μm
Lunghezza (sistema metrico) 10 mm
Diametro (sistema metrico) interno 2.1 mm
Fase stazionaria C18
Product Line Hypersil GOLD