Learn More
Thermo Scientific™ Thermo Scientific™ PCR Strip Cutting Tool
Product Code. 17869152 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17869152 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17869152 Supplier Thermo Scientific™ Supplier No. A55329
  • ONLINE EXCLUSIVE PRICE
    870.00€
    748.35€ / pezzi 1
    Risparmia 121.65€
    13% Sconto
 Restricted Item
Spedizione stimata: 24-11-2026
Easily and quickly cut the lysis and PCR tube strips in the SureTect workflow using the Thermo Scientific PCR Strip Cutting Tool.
Easily, quickly and safely cut the strip and separate either lysis or PCR tubes during the Thermo Scientific™ SureTect PCR Assay workflow using the Thermo Scientific™ PCR Strip Cutting Tool.
Designed specifically for use with the SureTect assays, the Thermo Scientific PCR Strip Cutting Tool is one in a suite of tools enabling a simple, streamlined workflow for maximum efficiency and productivity in your laboratory.
Use the Thermo Scientific PCR Strip Cutting Tool to quickly, safely and easily cut the strip and separate either lysis or PCR tubes into the desired number of reactions enabling flexible sample throughput and a fast, easy to use workflow
Quick and Easy
Use the Thermo Scientific PCR Strip Cutting Tool to speed the process of cutting the strip and seprate either lysis or PCR tubes into the desired number of assays. . The rugged material and design endures constant use so can be used time and time again whatever your sample throughput.
Combined with our comprehensive portfolio of assays and world-class instrumentation you can perform food safety tests confidently knowing exactly what you are getting – quality combined with simplicity.
We are committed to enabling you to experience the true confidence in your results, right-first time, every time, and the Thermo Scientific™ SureTect™ Real-Time PCR System is continually evolving to support your laboratory and testing needs.
TRUSTED_SUSTAINABILITY
Specifications
Product Type PCR Strip Cutting Tool
Description Thermo Scientific™ PCR Strip Cutting Tool
For Use With (Application) SureTect PCR Assay workflow
Quantity 1