Learn More
Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP
Product Code. 15332617 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
8 Reactions
Unit Size:
8 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15332617 8 Reactions 8 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15332617 Supplier Thermo Scientific™ Supplier No. 90407
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
In stock
Add to basket
Edge
Add to basket
Label primary antibodies with biotin for use in agarose or magnetic bead immunoprecipitation experiments using the Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP.
The Pierce Antibody Biotinylation Kit reagents have been optimized and validated to biotin label antibodies for IP and co-IP reactions. Determining the optimal number of biotins to attach to the target molecule is one of the major challenges of biotinylation. For IP and co-IP applications, too many biotins result in reduced affinity for the target antigen, while too few biotins result in antibody leaching upon elution of the target antigen. The biotin labeling procedure in the Pierce Antibody Biotinylation Kit for IP has been developed to address this challenge.
The Thermo Scientific™ Pierce™ Antibody Biotinylation Kit for IP provides biotinylation reagents designed specifically for the labeling of primary antibodies used in immunoprecipitation applications.
  • Optimized for IP - reagents and protocols developed specifically to address antibody labeling for immunoprecipitation applications
  • Easy to use - kit configuration specifically optimized to accommodate 50 to 200μg of antibody in a 100μL reaction; desalting columns are provided to remove excess biotinylation reagent after labeling
  • Amine reactive - reacts with primary amines (-NH2) of antibodies
  • Enhanced solubility - pegylation improves water solubility of the biotinylated antibody and prevents aggregation upon storage
  • Reduced steric hindrance - longer spacer arm (29 angstroms) minimize steric hindrance when binding to avidin molecules
The kit contains sufficient reagents to label 50 to 200μg of antibody in 100μL reaction volumes for eight samples. The NHS-PEG4-biotin labeling reagent contains an amine-reactive N-hydroxysuccinimide ester (NHS) group and a water-soluble PEG4 spacer for optimal labeling and is provided in easy-to-use, single-use microtubes. Both the labeling efficiency of the biotinylation reagent and binding affinity of the labeled antibody have been validated using mouse monoclonal (IgG1, IgG2), rabbit polyconal, and rabbit monoclonal antibodies. Zeba™ Desalting Spin Columns are provided for easy and efficient removal of salts and excess biotin.
Applications:
  • IP and co-IP
  • IP and co-IP for mass spectrometry applications
  • Antibody attachment to streptavidin agarose and magnetic supports
TRUSTED_SUSTAINABILITY
Specifications
Product Type Antibody Biotinylation Kit
Quantity 8 Reactions
Content And Storage EZ-Link NHS-PEG4-Biotin, 8 x 0.5 mg
Zeba Spin Desalting Columns (7K MWCO), 8 x 0.5 mL
20X Phosphate Buffered Saline, 15 mL
Store in refrigerator (2–8°C).
Labeling Scale 100 μg
Product Line Pierce
Label or Dye Biotin
Labeling Target Antibodies