Descrizione
- Extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions
- Amber vials available for light-sensitive compounds
- Supplied complete with Open-Top screw caps and PTFE/Rubber discs
Residue isolation, Derivative preparation, Maximum sample retrieval, Moisture protection, Sample storage, Precipitations, Centrifugations, Solvent evaporation
Specifica
Specifica
| Colore | Trasparente |
| Materiale | Vetro di silice |
| Volume (sistema metrico) | 5 ml |
| Stile filo | Interno |
| Volume (sistema metrico) totale | 5 mL |
| Tipo di chiusura | Tappo a vite |
| Certificazione/Conformità | Certificate of Analysis (COA) |
| Product Type | Vial |